Pramiracetam hydrate 98%

Supplier: Ambeed
Warning

Synonyms: N-(2-(Diisopropylamino)ethyl)-2-(2-oxopyrrolidin-1-yl)acetamide

A241738-1G A241738-1MG A241738-250MG A241738-25MG A241738-50MG A241738-5G A241738-5MG
77238-098EA 62.83 USD
77238-098 77238-100 77238-102 77238-104 77238-106 77238-108 77238-110
Pramiracetam hydrate 98%
Pramiracetam hydrate

Precaución: for Research Use Only

Formula: C₁₄H₂₇N₃O₂
MW: 269.39 g/mol
MDL Number: MFCD00867219
CAS Number: 68497-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR