tert-Butyl (2-((tert-butoxycarbonyl)amino)ethyl)(2-hydroxyethyl)carbamate 97%

Supplier: Ambeed
Warning

A241493-100MG A241493-1G A241493-250MG
77238-096EA 55 USD
77238-096 77593-544 77593-546
tert-Butyl (2-((tert-butoxycarbonyl)amino)ethyl)(2-hydroxyethyl)carbamate 97%
tert-Butyl (2-((tert-butoxycarbonyl)amino)ethyl)(2-hydroxyethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₄H₂₈N₂O₅
MDL Number: MFCD24466012
CAS Number: 200283-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR