9H-fluorene-2,7-diol 97%

Supplier: Ambeed
Warning

A241457-100MG A241457-250MG A241457-5G
77249-780EA 17 USD
77249-780 77249-782 77249-784
9H-fluorene-2,7-diol 97%
9H-fluorene-2,7-diol

Precaución: for Research Use Only

Formula: C₁₃H₁₀O₂
MDL Number: MFCD15144693
CAS Number: 5043-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR