Ethyl 2-(hydroxymethyl)nicotinate 95%

Supplier: Ambeed
Warning

A241436-100MG A241436-1G A241436-250MG
77261-800EA 57.68 USD
77261-800 77261-802 77261-804
Ethyl 2-(hydroxymethyl)nicotinate 95%
Ethyl 2-(hydroxymethyl)nicotinate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.18 g/mol
MDL Number: MFCD30067507
CAS Number: 1346539-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR