6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole 97%

Supplier: Ambeed
Warning

A241346-250MG A241346-10G
77276-016EA 6 USD
77276-016 77568-344
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole 97%
6-Fluoro-3-(4-piperidinyl)-1,2-benzisoxazole

Precaución: For research use only.

Formula: C₁₂H₁₃FN₂O
MW: 220.24 g/mol
MDL Number: MFCD03236724
CAS Number: 84163-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR