2,2'-Bipyridine-3,3'-dicarboxylic acid 97%

Supplier: Ambeed
Warning

A241343-1G A241343-25G A241343-5G
77238-090EA 9 USD
77238-090 77238-092 77238-094
2,2'-Bipyridine-3,3'-dicarboxylic acid 97%
2,2'-Bipyridine-3,3'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₈N₂O₄
MDL Number: MFCD00226068
CAS Number: 4433-01-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR