N-(3-((2-((3-Fluoro-4-(4-methylpiperazin-1-yl)phenyl)amino)-7H-pyrrolo[2,3-d]pyrimidin-4-yl)oxy)phenyl)acrylamide 97%

Supplier: Ambeed
Warning

Synonyms: Avitinib

A241112-100MG A241112-1MG A241112-25MG A241112-5MG
77238-082EA 242.05 USD
77238-082 77238-084 77238-086 77238-088
N-(3-((2-((3-Fluoro-4-(4-methylpiperazin-1-yl)phenyl)amino)-7H-pyrrolo[2,3-d]pyrimidin-4-yl)oxy)phenyl)acrylamide 97%
N-(3-((2-((3-Fluoro-4-(4-methylpiperazin-1-yl)phenyl)amino)-7H-pyrrolo[2,3-d]pyrimidin-4-yl)oxy)phenyl)acrylamide

Precaución: for Research Use Only

Formula: C₂₆H₂₆FN₇O₂
MW: 487.53 g/mol
MDL Number: MFCD29089376
CAS Number: 1557267-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR