(S)-Phanephos 98%

Supplier: Ambeed
Warning

Synonyms: (S)-(+)-4,12-Bis(diphenylphosphino)-[2.2]-paracyclophane, (S)-PHANEPHOS, (S)-4,12-Bis(diphenylphosphino)-[2.2]paracyclophane

A241059-100MG A241059-250MG A241059-50MG
77249-768EA 114.33 USD
77249-768 77249-770 77249-772
(S)-Phanephos 98%
(S)-Phanephos

Precaución: for Research Use Only

Formula: C₄₀H₃₄P₂
MW: 576.65 g/mol
MDL Number: MFCD03094575
CAS Number: 192463-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR