2-Tetrahydro-2H-pyran-4-ylethanol 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Tetrahydro-2H-pyran-4-yl)ethanol, 4-(2-Hydroxyethyl)tetrahydro-2H-pyran

A241028-10G A241028-1G A241028-5G A241028-250MG
77249-762EA 334.75 USD
77249-762 77249-764 77249-766 77746-361
2-Tetrahydro-2H-pyran-4-ylethanol 98%
2-Tetrahydro-2H-pyran-4-ylethanol

Precaución: for Research Use Only

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
MDL Number: MFCD00129068
CAS Number: 4677-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR