3-(4-Chlorophenyl)propionic acid 98%

Supplier: Ambeed
Warning

A240974-100G A240974-10G A240974-25G A240974-5G
77276-006EA 81 USD
77276-006 77276-008 77276-010 77276-012 77276-014
3-(4-Chlorophenyl)propionic acid 98%
3-(4-Chlorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₂
MDL Number: MFCD00016555
CAS Number: 2019-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR