2-Amino-4-chlorothiophenol 95%

Supplier: Ambeed
Danger

A240949-25G A240949-5G
77249-758EA 26 USD
77249-758 77249-760
2-Amino-4-chlorothiophenol 95%
2-Amino-4-chlorothiophenol

Precaución: for Research Use Only

Formula: C₆H₆ClNS
MDL Number: MFCD00792528
CAS Number: 1004-00-8
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR