2-(Pyridin-3-yl)aniline 95%

Supplier: Ambeed
Warning

Synonyms: 3-(2-Aminophenyl)pyridine

A240885-1G A240885-250MG
77276-002EA 162.74 USD
77276-002 77276-004
2-(Pyridin-3-yl)aniline 95%
2-(Pyridin-3-yl)aniline

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂
MW: 170.21 g/mol
MDL Number: MFCD08060516
CAS Number: 177202-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR