4-Chlorobutyl acetate 98%

Supplier: Ambeed
Warning

A240851-100G A240851-10G A240851-25G A240851-500G A240851-5G
77288-718EA 27 USD
77288-718 77288-720 77288-722 77288-724 77288-726
4-Chlorobutyl acetate 98%
4-Chlorobutyl acetate

Precaución: for Research Use Only

Formula: C₆H₁₁ClO₂
MDL Number: MFCD00001013
CAS Number: 6962-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR