CID-5721353 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(5-(5-Bromo-2-oxoindolin-3-ylidene)-4-oxo-2-thioxothiazolidin-3-yl)succinic acid, 2-((5Z)-5-(5-Bromo-2-oxo-1H-indol-3-ylidene)-4-oxo-2-sulphanylidene-1,3-thiazolidin-3-yl)butanedioic acid
A240729-10MG
A240729-1MG
A240729-25MG
A240729-5MG
77261-790EA
87
USD
77261-790
77261-792
77261-794
77261-796
CID-5721353 97%
CID-5721353
Precaución: for Research Use Only
Formula:
C₁₅H₉BrN₂O₆S₂ MW: 457.23 g/mol |
MDL Number:
MFCD00366372 CAS Number: 301356-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...