SR1001 98%

Supplier: Ambeed
Warning

Synonyms: N-(5-(N-(4-(1,1,1,3,3,3-hexafluoro-2-hydroxypropan-2-yl)phenyl)sulphamoyl)-4-methylthiazol-2-yl)acetamide

A240693-10MG A240693-1MG A240693-50MG A240693-5MG
77238-074EA 22.66 USD
77238-074 77238-076 77238-078 77238-080
SR1001 98%
SR1001

Precaución: for Research Use Only

Formula: C₁₅H₁₃F₆N₃O₄S₂
MW: 477.40 g/mol
MDL Number: MFCD23160036
CAS Number: 1335106-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR