Zinc 2-Mercaptobenzothiazole 97+%

Supplier: Ambeed
Danger

Synonyms: 2-Mercaptobenzothiazolezincsalt, 2-Mercaptobenzothiazole Zinc Salt, Zinc 2-Benzothiazolethiolate

A240625-25G A240625-100G A240625-500G
77261-780EA 8 USD
77261-780 77615-986 77615-988
Zinc 2-Mercaptobenzothiazole 97+%
Zinc 2-Mercaptobenzothiazole

Precaución: for Research Use Only

Formula: C₁₄H₈N₂S₄Zn
MW: 397.85 g/mol
MDL Number: MFCD00072234
CAS Number: 155-04-4
UN: 3077
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR