(2-Amino-5-methylphenyl)phosphonic acid 98%

Supplier: Ambeed
Warning

A240240-1G A240240-250MG
77249-748EA 594 USD
77249-748 77249-750
(2-Amino-5-methylphenyl)phosphonic acid 98%
(2-Amino-5-methylphenyl)phosphonic acid

Precaución: for Research Use Only

Formula: C₇H₁₀NO₃P
MDL Number: MFCD20699745
CAS Number: 69675-98-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR