5-Nitropyridin-3-ol 98%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-5-nitropyridine

A240147-100MG A240147-1G A240147-250MG
77275-996EA 37 USD
77275-996 77275-998 77276-000
5-Nitropyridin-3-ol 98%
5-Nitropyridin-3-ol

Precaución: for Research Use Only

Formula: C5H4N2O3
MDL Number: MFCD04114224
CAS Number: 186593-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR