2-Bromo-9,9-dihexylfluorene 97%

Supplier: Ambeed
Warning

A239176-1G A239176-25G A239176-5G
77238-066EA 8 USD
77238-066 77238-068 77238-070 77238-072
2-Bromo-9,9-dihexylfluorene 97%
2-Bromo-9,9-dihexylfluorene

Precaución: for Research Use Only

Formula: C₂₅H₃₃Br
MDL Number: MFCD14584649
CAS Number: 226070-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR