4-(2,5-Dimethylphenyl)-4-oxobutanoic acid 97%

Supplier: Ambeed
Warning

A239158-100MG A239158-250MG
77288-706EA 52 USD
77288-706 77288-708
4-(2,5-Dimethylphenyl)-4-oxobutanoic acid 97%
4-(2,5-Dimethylphenyl)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄O₃
MDL Number: MFCD00020537
CAS Number: 5394-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR