4,4'-Dithiodimorpholine 98%

Supplier: Ambeed
Warning

Synonyms: N,N'-Dithiobis(morpholine), 4-(4-Morpholinyldisulphanyl)morpholine, N,N'-Dimorpholine Disulphide

A239147-100G A239147-25G A239147-500G
77288-700EA 17.51 USD
77288-700 77288-702 77288-704
4,4'-Dithiodimorpholine 98%
4,4'-Dithiodimorpholine

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₂S₂
MW: 236.35 g/mol
MDL Number: MFCD00023319
CAS Number: 103-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR