N-(tert-Butoxycarbonyl)-L-aspartic acid 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-L-aspartic Acid, Boc-Asp-OH

A239123-25G A239123-500G
77275-968EA 13 USD
77275-968 77275-970 77748-414
N-(tert-Butoxycarbonyl)-L-aspartic acid 98%
N-(tert-Butoxycarbonyl)-L-aspartic acid

Precaución: for Research Use Only

Formula: C₉H₁₅NO₆
MW: 233.22 g/mol
MDL Number: MFCD00037279
CAS Number: 13726-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR