4-(Methylsulfonyl)-2-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A238986-1000G A238986-100G A238986-10G A238986-25G A238986-500G A238986-5G
77238-054EA 84 USD
77238-054 77238-056 77238-058 77238-060 77238-062 77238-064
4-(Methylsulfonyl)-2-nitrobenzoic acid 98%
4-(Methylsulphonyl)-2-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₆S
MDL Number: MFCD01941299
CAS Number: 110964-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR