4-Bromo-3-fluoroaniline 98%

Supplier: Ambeed
Warning

A238640-100G A238640-10G A238640-1G A238640-25G A238640-500G A238640-5G
77288-676EA 55 USD
77288-676 77288-678 77288-680 77288-682 77288-684 77288-686
4-Bromo-3-fluoroaniline 98%
4-Bromo-3-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrFN
MDL Number: MFCD00672933
CAS Number: 656-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR