Zonisamide 98%

Supplier: Ambeed
Warning

A238577-1MG A238577-250MG A238577-50MG A238577-5G A238577-1G
77249-732EA 7 USD
77249-732 77249-734 77249-736 77615-970 77746-356
Zonisamide 98%
Zonisamide

Precaución: for Research Use Only

Formula: C₈H₈N₂O₃S
MDL Number: MFCD00865316
CAS Number: 68291-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR