4-Bromocinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Bromophenyl)acrylic acid

A238540-100G A238540-10G A238540-25G A238540-500G A238540-5G
77261-724EA 43.26 USD
77261-724 77261-726 77261-728 77261-730 77261-732
4-Bromocinnamic acid 98%
4-Bromocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₇BrO₂
MW: 227.06 g/mol
MDL Number: MFCD00004394
CAS Number: 1200-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR