3-(2,4-Dichlorophenyl)propionic acid 98%

Supplier: Ambeed
Danger

Synonyms: 3-(2,4-Dichlorophenyl)propanoic acid

A238353-1G A238353-25G A238353-5G
77261-718EA 11.33 USD
77261-718 77261-720 77261-722
3-(2,4-Dichlorophenyl)propionic acid 98%
3-(2,4-Dichlorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₂
MW: 219.07 g/mol
MDL Number: MFCD00275605
CAS Number: 55144-92-8
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR