(S)-2-(6-Benzylpyridin-2-yl)-4-(tert-butyl)-4,5-dihydrooxazole 98%
Supplier: Ambeed
|
Warning
|
A238044-100MG
A238044-1G
A238044-250MG
77261-702EA
129.78
USD
77261-702
77261-704
77261-706
(S)-2-(6-Benzylpyridin-2-yl)-4-(tert-butyl)-4,5-dihydrooxazole 98%
(S)-2-(6-Benzylpyridin-2-yl)-4-(tert-butyl)-4,5-dihydrooxazole
Precaución: for Research Use Only
Formula:
C₁₉H₂₂N₂O MW: 294.39 g/mol |
CAS Number:
1800190-39-9 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...