4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)-4-oxobutanoic acid 97%

Supplier: Ambeed
Warning

A237997-1G A237997-5G A237997-250MG
77288-650EA 57.68 USD
77288-650 77288-652 77615-962
4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)-4-oxobutanoic acid 97%
4-(4-(tert-Butoxycarbonyl)piperazin-1-yl)-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₂O₅
MW: 286.32 g/mol
MDL Number: MFCD06409184
CAS Number: 288851-44-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR