Benzoin isopropyl ether 97%

Supplier: Ambeed
Warning

A237894-1000G A237894-100G A237894-25G A237894-500G
77288-642EA 102 USD
77288-642 77288-644 77288-646 77288-648
Benzoin isopropyl ether 97%
Benzoin isopropyl ether

Precaución: for Research Use Only

Formula: C₁₇H₁₈O₂
MDL Number: MFCD00015025
CAS Number: 6652-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR