(1S)-3,3'-Dimethyl-[1,1'-binaphthalene]-2,2'-diol 98%

Supplier: Ambeed
Warning

A237816-100MG A237816-1G A237816-250MG
77249-698EA 99.91 USD
77249-698 77249-700 77249-702
(1S)-3,3'-Dimethyl-[1,1'-binaphthalene]-2,2'-diol 98%
(1S)-3,3'-Dimethyl-[1,1'-binaphthalene]-2,2'-diol

Precaución: for Research Use Only

Formula: C₂₂H₁₈O₂
MW: 314.37 g/mol
MDL Number: MFCD00187966
CAS Number: 55515-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR