2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium chloride 98%

Supplier: Ambeed
Warning

Synonyms: SEM-triphenylphosphonium Chloride, Triphenyl((2-(trimethylsilyl)ethoxy)methyl)phosphonium chloride

A237762-1G A237762-5G A237762-25G
77249-694EA 19.57 USD
77249-694 77249-696 77593-526
2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium chloride 98%
2-(Trimethylsilyl)ethoxymethyltriphenylphosphonium chloride

Precaución: for Research Use Only

Formula: C₂₄H₃₀ClOPSi
MW: 429.01 g/mol
MDL Number: MFCD00043159
CAS Number: 82495-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR