2-Chloro-3,5-diiodopyridine 97%

Supplier: Ambeed
Warning

A237561-100MG A237561-1G
77261-674EA 7.21 USD
77261-674 77261-676
2-Chloro-3,5-diiodopyridine 97%
2-Chloro-3,5-diiodopyridine

Precaución: for Research Use Only

Formula: C₅H₂ClI₂N
MW: 365.34 g/mol
MDL Number: MFCD04966987
CAS Number: 856169-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR