N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4,6-dichloro-5-formamidopyrimidine, N-(2-Amino-4,6-dichloro-5-pyrimidinyl)formamide

A237451-100G A237451-10G A237451-25G A237451-500G A237451-5G
77288-622EA 38 USD
77288-622 77288-624 77288-626 77288-628 77288-630
N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide 98% (by HPLC)
N-(2-Amino-4,6-dichloropyrimidine-5-yl)formamide

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₄O
MDL Number: MFCD04112936
CAS Number: 171887-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR