2-Decanone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Decan-2-one
A237354-100G
A237354-25G
A237354-500G
A237354-5G
77261-666EA
122.57
USD
77261-666
77261-668
77261-670
77261-672
2-Decanone 98%
2-Decanone
Precaución: for Research Use Only
Formula:
C₁₀H₂₀O MW: 156.27 g/mol |
MDL Number:
MFCD00009571 CAS Number: 693-54-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...