(R)-Ethyl 5-([1,1'-biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpent-2-enoate 97%
Supplier: Ambeed
|
Warning
|
A237269-1G
A237269-250MG
A237269-5G
77249-676EA
5.15
USD
77249-676
77249-678
77249-680
77249-682
(R)-Ethyl 5-([1,1'-biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpent-2-enoate 97%
(R)-Ethyl 5-([1,1'-biphenyl]-4-yl)-4-((tert-butoxycarbonyl)amino)-2-methylpent-2-enoate
Precaución: for Research Use Only
Formula:
C₂₅H₃₁NO₄ |
MDL Number:
MFCD23378892 CAS Number: 149709-59-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...