4-Ethoxyphenyl trans-4-Propylcyclohexanecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Trans-4-ethoxyphenyl 4-propylcyclohexanecarboxylate, trans-4-Propylcyclohexanecarboxylic acid 4-Ethoxyphenyl ester

A237111-10G A237111-25G A237111-5G
77288-610EA 25.75 USD
77288-610 77288-612 77288-614
4-Ethoxyphenyl trans-4-Propylcyclohexanecarboxylate 97%
4-Ethoxyphenyl trans-4-Propylcyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₁₈H₂₆O₃
MW: 290.40 g/mol
MDL Number: MFCD11053417
CAS Number: 67589-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR