3,5-Bis(ethoxycarbonyl)-2,6-dimethyl-1,4-dihydropyridine-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A236906-100MG A236906-1G A236906-250MG A236906-5G
77249-666EA 39.14 USD
77249-666 77249-668 77249-670 77249-672
3,5-Bis(ethoxycarbonyl)-2,6-dimethyl-1,4-dihydropyridine-4-carboxylic acid 97%
3,5-Bis(ethoxycarbonyl)-2,6-dimethyl-1,4-dihydropyridine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₆
MW: 297.30 g/mol
MDL Number: MFCD00417306
CAS Number: 19350-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR