Letermovir 98%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(8-Fluoro-3-(2-methoxy-5-(trifluoromethyl)phenyl)-2-(4-(3-methoxyphenyl)piperazin-1-yl)-3,4-dihydroquinazolin-4-yl)acetic acid

A236700-100MG A236700-1MG A236700-50MG A236700-5MG A236700-10MG
77249-658EA 221.45 USD
77249-658 77249-660 77249-662 77249-664 77615-954
Letermovir 98%
Letermovir

Precaución: for Research Use Only

Formula: C₂₉H₂₈F₄N₄O₄
MW: 572.55 g/mol
MDL Number: MFCD22572725
CAS Number: 917389-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR