(R)-1-(3-Fluoro-4-(trifluoromethyl)phenyl)ethanamine 97%

Supplier: Ambeed
Warning

A236694-100MG A236694-1G A236694-250MG
77261-660EA 48.41 USD
77261-660 77261-662 77261-664
(R)-1-(3-Fluoro-4-(trifluoromethyl)phenyl)ethanamine 97%
(R)-1-(3-Fluoro-4-(trifluoromethyl)phenyl)ethanamine

Precaución: for Research Use Only

Formula: C₉H₉F₄N
MW: 207.16 g/mol
MDL Number: MFCD06762190
CAS Number: 1079656-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR