Acetrizoic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Acetamido-2,4,6-triiodobenzoic Acid

A236609-100MG A236609-25MG A236609-1MG
77261-656EA 13.39 USD
77261-656 77261-658 77615-952
Acetrizoic acid 98%
Acetrizoic acid

Precaución: for Research Use Only

Formula: C₉H₆I₃NO₃
MW: 556.86 g/mol
MDL Number: MFCD00016495
CAS Number: 85-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR