5-Methyl-2H-benzo[d][1,3]oxazine-2,4(1H)-dione 97%

Supplier: Ambeed
Warning

A236486-100MG A236486-1G A236486-250MG
77261-650EA 53.56 USD
77261-650 77261-652 77261-654
5-Methyl-2H-benzo[d][1,3]oxazine-2,4(1H)-dione 97%
5-Methyl-2H-benzo[d][1,3]oxazine-2,4(1H)-dione

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.16 g/mol
MDL Number: MFCD00797331
CAS Number: 20877-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR