4,4'-Bis(dimethylamino)benzil 99%

Supplier: Ambeed
Warning

Synonyms: 1,2-Bis(4-(dimethylamino)phenyl)ethane-1,2-dione, 1,2-Bis(4-dimethylaminophenyl)-1,2-ethanedione

A236428-1G A236428-250MG A236428-5G
77288-594EA 8.24 USD
77288-594 77288-596 77288-598 77288-600
4,4'-Bis(dimethylamino)benzil 99%
4,4'-Bis(dimethylamino)benzil

Precaución: for Research Use Only

Formula: C₁₈H₂₀N₂O₂
MDL Number: MFCD00051389
CAS Number: 17078-27-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR