2H-1,4-Benzothiazin-3(4H)-one 98%

Supplier: Ambeed
Warning

A236331-100G A236331-1G A236331-25G A236331-5G
77238-010EA 165 USD
77238-010 77238-012 77238-014 77238-016
2H-1,4-Benzothiazin-3(4H)-one 98%
2H-1,4-Benzothiazin-3(4H)-one

Precaución: for Research Use Only

Formula: C₈H₇NOS
MDL Number: MFCD00006698
CAS Number: 5325-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR