1-Bromo-3-fluoro-4-iodobenzene 98%

Supplier: Ambeed
Warning

A236238-100G A236238-10G A236238-25G A236238-500G A236238-5G
77288-578EA 35 USD
77288-578 77288-580 77288-582 77288-584 77288-586
1-Bromo-3-fluoro-4-iodobenzene 98%
1-Bromo-3-fluoro-4-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFI
MDL Number: MFCD00010608
CAS Number: 105931-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR