Azadibenzocyclooctyne-amine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
DBCO-amine, 3-Amino-1-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)-1-propanone, Dibenzocyclooctyne-amine
A236148-100MG
A236148-1G
A236148-50MG
A236148-250MG
A236148-5G
77261-646EA
399.64
USD
77261-646
77261-648
77615-942
77615-940
77748-410
Azadibenzocyclooctyne-amine 98%
Azadibenzocyclooctyne-amine
Precaución: for Research Use Only
Formula:
C₁₈H₁₆N₂O MW: 276.33 g/mol |
MDL Number:
MFCD22380759 CAS Number: 1255942-06-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...