3,3,3-Tris(4-chlorophenyl)propionic acid 97%

Supplier: Ambeed
Warning

A235985-1G A235985-25G A235985-5G
77275-908EA 7 USD
77275-908 77275-910 77275-912
3,3,3-Tris(4-chlorophenyl)propionic acid 97%
3,3,3-Tris(4-chlorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₅Cl₃O₂
MDL Number: MFCD00010264
CAS Number: 2168-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR