IM-12 98%

Supplier: Ambeed
Warning

Synonyms: 3-((4-Fluorophenethyl)amino)-1-methyl-4-(2-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione

A235864-100MG A235864-10MG A235864-1MG A235864-50MG A235864-5MG
77237-996EA 295.61 USD
77237-996 77237-998 77238-000 77238-002 77238-004
IM-12 98%
IM-12

Precaución: for Research Use Only

Formula: C₂₂H₂₀FN₃O₂
MW: 377.4115 g/mol
MDL Number: MFCD20527313
CAS Number: 1129669-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR