5-Nitro-m-toluidine 98%

Supplier: Ambeed
Warning

A235862-10G A235862-1G A235862-250MG A235862-5G
77275-900EA 95 USD
77275-900 77275-902 77275-904 77275-906
5-Nitro-m-toluidine 98%
5-Nitro-m-toluidine

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MDL Number: MFCD00082655
CAS Number: 618-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR