3-Chloro-6-nitro-1H-indazole 97%

Supplier: Ambeed
Warning

A235660-100G A235660-100MG A235660-1G A235660-250MG A235660-25G A235660-5G
77275-880EA 1153.6 USD
77275-880 77275-882 77275-884 77275-886 77275-888 77275-890
3-Chloro-6-nitro-1H-indazole 97%
3-Chloro-6-nitro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄ClN₃O₂
MW: 197.5800 g/mol
MDL Number: MFCD00010741
CAS Number: 50593-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR